missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctanesulfonamide, 95%

Product Code. 15549493 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15549493 1 g 1g
15559493 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15549493 Supplier Thermo Scientific Acros Supplier No. 459640010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 754-91-6 | C8H2F17NO2S | 499.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-91-6
Molecular Formula C8H2F17NO2S
Molecular Weight (g/mol) 499.15
InChI Key RRRXPPIDPYTNJG-UHFFFAOYSA-N
Synonym perfluorooctanesulfonamide, perfluoroctylsulfonamide, perfluorooctane sulfonamide, pfosa, heptadecafluorooctanesulphonamide, perfluorooctanesulfonic acid amide, unii-80am718fml, 1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro, deethylsulfluramie, perfluorooctanesulphonamide
PubChem CID 69785
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide
SMILES C(C(C(C(C(F)(F)S(=O)(=O)N)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 154.0°C to 155.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (mix of isomers) (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 499.15
Percent Purity 95%
Chemical Name or Material Perfluorooctanesulfonamide
compliance-icons compliance-icons
Product Identifier
  • Perfluorooctanesulfonamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.