missing translation for 'onlineSavingsMsg'
Learn More

Perfluorononanoic acid, 97%

Product Code. 11499060
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499060 5 g 5g
11409070 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11499060 Supplier Thermo Scientific Alfa Aesar Supplier No. A13980.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 375-95-1 | C9HF17O2 | 464.078 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-95-1
Molecular Formula C9HF17O2
Molecular Weight (g/mol) 464.078
MDL Number MFCD00039605
InChI Key UZUFPBIDKMEQEQ-UHFFFAOYSA-N
Synonym perfluorononanoic acid, heptadecafluorononanoic acid, perfluoro-n-nonanoic acid, nonanoic acid, heptadecafluoro, perfluorononan-1-oic acid, pfna, heptadecafluoropelargonic acid, perfluornonansaeure, heptadecafluornonansaeure, heptadekafluornonansaeure
PubChem CID 67821
ChEBI CHEBI:38397
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 63°C to 68°C
Boiling Point 218°C
Linear Formula CF3(CF2)7CO2H
Quantity 5 g
UN Number UN3261
Beilstein 1897287
Formula Weight 464.07
Percent Purity 97%
Chemical Name or Material Perfluorononanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.