missing translation for 'onlineSavingsMsg'
Learn More

Perfluorokerosene (PFK), high boiling, 50% in PFK low boiling, for mass spectrometry

Product Code. 13415122 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13415122 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13415122 Supplier Thermo Scientific Acros Supplier No. 450130010
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.8900g/mL
Boiling Point <180°C
Flash Point 46°C
Packaging Glass bottle
Linear Formula CnF2n+2
Quantity 1 mL
Specific Gravity 1.89
Grade For mass spectrometry
Chemical Name or Material Perfluorokerosene (PFK), high boiling, 50% in PFK low boiling
compliance-icons
Product Identifier
  • Perfluorokerosene, high boiling, 50% in Perfluorokerosene low boiling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.