missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexyl Iodide, TRC

Product Code. 30411549
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411549 1 g -
30419306 25 g -
30424744 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411549 Supplier Toronto Research Chemicals Supplier No. TRCP2275551G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-43-1
Molecular Formula C6F13I
Molecular Weight (g/mol) 445.95
Synonym 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-6-iodohexane, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-iodohexane, 1-Iodo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane, 1-Iodofluorohexane, 1-Iodoperfluorohexane, 1-Iodotridecafluorohexane, 6-Iodoperfluorohexane, Perfluoro-1-iodohexane, Perfluorohexyl iodide, Tridecafluoro-1-iodohexane, Tridecafluoro-6-iodohexane, Tridecafluorohexyl iodide, n-Perfluorohexyl iodide
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6F13I/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)20
Formula Weight 445.8837
Chemical Name or Material Perfluorohexyl Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.