missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexyl bromide, 97%

Product Code. 15520624 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15520624 25 g 25g
15510624 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15520624 Supplier Thermo Scientific Acros Supplier No. 459310250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 335-56-8 | C6BrF13 | 398.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-56-8
Molecular Formula C6BrF13
Molecular Weight (g/mol) 398.95
InChI Key JTYRBFORUCBNHJ-UHFFFAOYSA-N
Synonym 1-bromoperfluorohexane, perfluorohexyl bromide, perfluorohexylbromide, tridecafluorohexyl bromide, 1-bromo-tridecafluorohexane, bromoperfluorohexane, acmc-1cmhf, 1-bromotridecafluorohexane, tridecafluoro-1-bromohexane
PubChem CID 92755
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
SMILES C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.8710g/mL
Boiling Point 98.0°C to 100.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula F3C(CF2)4CF2Br
Quantity 25 g
Specific Gravity 1.871
Formula Weight 398.95
Percent Purity 97%
Chemical Name or Material Perfluorohexyl bromide
compliance-icons
Product Identifier
  • Perfluorohexyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.