missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluorohexanes (Mixture of Isomers), TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Perfluorohexanes (Mixture of Isomers) |
| CAS | 355-42-0 |
| Molecular Formula | C6F14 |
| Quantity | 500 mg |
| Synonym | Tetradecafluorohexane, Hexane','1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluoro-, Hexane','tetradecafluoro- (6CI,8CI,9CI), 1,1,1,2,2,3,3,4,4,5,5,6,6,6-Tetradecafluorohexane, AF 0150, FC 72, Fluorinert FC 72, Fluorinert FX 3250D, Fluorinert PF 5060, Flutec PP 1, PF 5060, PFC 51-14, PP 1, Perflexane, Perfluoro-n-hexane, Perfluorohexane, Tetradecafluorohexane, n-Tetradecafluorohexane |
| InChI Formula | InChI=1S/C6F14/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)20 |
| SMILES | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane |
| Molecular Weight (g/mol) | 338.04 |
| Formula Weight | 337.9776 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction