missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexanes, 98+%, Thermo Scientific Chemicals

Product Code. 11490480
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490480 500 g 500g
11327148 25 g 25g
11317148 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11490480 Supplier Thermo Scientific Alfa Aesar Supplier No. 015812.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Perfluorohexanes is used as a coolant, photosensitizer in fluorous biphasic singlet oxygenation and an inert reaction medium. It is used in the electronic cooling liquid and insulator. It plays a vital role as a reaction medium, especially for photooxidation reactions. Further, it finds application as an ultrasound contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-42-0
Molecular Formula C6F14
Molecular Weight (g/mol) 338.044
MDL Number MFCD00000437
InChI Key ZJIJAJXFLBMLCK-UHFFFAOYSA-N
Synonym perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn
PubChem CID 9639
ChEBI CHEBI:39427
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point -4°C
Color Colorless
Density 1.699 g/mL
Boiling Point 58°C to 60°C
Flash Point None
Linear Formula CF3(CF2)4CF3
Refractive Index 1.251
Quantity 500 g
Beilstein 1802113
Solubility Information Immiscible with water.
Formula Weight 338.04
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Perfluorohexanes
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.