missing translation for 'onlineSavingsMsg'
Learn More

Perfluoroheptanoic Acid, TRC

Product Code. 18091340
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18091340 5 g 5g
18004050 1 g 1g
18052250 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18091340 Supplier Toronto Research Chemicals Supplier No. TRCP2863005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-85-9
Molecular Formula C7 H F13 O2
Molecular Weight (g/mol) 364.06
Synonym Heptanoic acid, tridecafluoro- (6CI,8CI,9CI), 2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptanoic acid, PFHpA, Perfluoro-n-heptanoic acid, Perfluoroenanthic acid, Perfluoroheptanoic acid, Tridecafluoroheptanoic acid, PFHpA
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7HF13O2/c8-2(9,1(21)22)3(10,11)4(12,13)5(14,15)6(16,17)7(18,19)20/h(H,21,22)
Formula Weight 363.9769
Percent Purity >95%
Chemical Name or Material Perfluoroheptanoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.