missing translation for 'onlineSavingsMsg'
Learn More

Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals

Product Code. 11318985
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318985 5 g 5g
11308985 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11318985 Supplier Thermo Scientific Alfa Aesar Supplier No. L14113.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 307-55-1 | Molecular Formula: C12HF23O2 | Molecular Weight: 614.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-55-1
Molecular Formula C12HF23O2
Molecular Weight (g/mol) 614.101
MDL Number MFCD00198081
InChI Key CXGONMQFMIYUJR-UHFFFAOYSA-N
Synonym perfluorododecanoic acid, tricosafluorododecanoic acid, perfluorolauric acid, dodecanoic acid, tricosafluoro, unii-p6g1j7nm2d, p6g1j7nm2d, pfdoa, tricosafluorolauric acid, n-perfluorododecanoic acid, n-c11f23cooh
PubChem CID 67545
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 108°C to 112°C
Boiling Point 245°C
Flash Point None
Linear Formula CF3(CF2)10CO2H
Quantity 5 g
UN Number UN3261
Beilstein 1811257
Formula Weight 614.1
Percent Purity 96%
Chemical Name or Material Perfluorododecanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.