missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Perfluorodecalin, TRC
Tekniske data
Tekniske data
| Chemical Name or Material | Perfluorodecalin |
| CAS | 306-94-5 |
| Molecular Formula | C10F18 |
| Quantity | 50 g |
| Synonym | 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-Octadecafluorodecahydronaphthalene, APF 140, Decalin Perfluoride, FDC, Flutec PP 5, Flutec PP 6, Flutec PP 7, NSC 97066, Octadecafluorodecahydronaphthalene, Octadecafluorodecalin, PP 5, PP 6, Perflunafene, Perfluorodecahydronaphthalene, Perfluorodecalin |
| InChI Formula | InChI=1S/C10F18/c11-1-2(12,5(17,18)9(25,26)7(21,22)3(1,13)14)6(19,20)10(27,28)8(23,24)4(1,15)16 |
| SMILES | FC1(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C1(F)F |
| IUPAC Name | 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene |
| Molecular Weight (g/mol) | 462.08 |
| Formula Weight | 461.9713 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?