missing translation for 'onlineSavingsMsg'
Learn More

Perfluorobutyl Iodide, TRC

Product Code. 30427225
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427225 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427225 Supplier Toronto Research Chemicals Supplier No. TRCP2862055G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluorobutyl Iodide
CAS 423-39-2
Purity Grade Notes GC
Molecular Formula C4F9I
Quantity 5 g
Synonym Perfluoro-n-butyl Iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, 1-Iodoperfluorobutane, 4-Iodoperfluorobutane, Nonafluoro-1-iodobutane
InChI Formula InChI=1S/C4F9I/c5-1(6,3(9,10)11)2(7,8)4(12,13)14
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)I
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
Molecular Weight (g/mol) 345.93
Formula Weight 345.8901
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.