missing translation for 'onlineSavingsMsg'
Learn More

Perfluorobutyl iodide, 99%

Product Code. 10075723 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10075723 100 g Glass bottle 100g
10389050 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10075723 Supplier Thermo Scientific Acros Supplier No. 269951000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 423-39-2 | C4F9I | 345.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-39-2
Molecular Formula C4F9I
Molecular Weight (g/mol) 345.92
MDL Number MFCD00001062
InChI Key PGRFXXCKHGIFSV-UHFFFAOYSA-N
Synonym perfluorobutyl iodide, nonafluoro-1-iodobutane, nonafluorobutyl iodide, perfluorobutyliodide, n-nonafluorobutyl iodide, perfluoro-n-butyl iodide, 1-iodoperfluorobutane, ccris 9008, 1-iodononafluorobutane, butane, 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodo
PubChem CID 67917
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
SMILES C(C(C(F)(F)I)(F)F)(C(F)(F)F)(F)F

Specifications

Melting Point -88°C
Color Purple
Density 2.0100g/mL
Boiling Point 64°C to 66°C (973.0 mbar)
Flash Point >115°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3(CF2)3I
Refractive Index 1.3275 to 1.3295
Quantity 100 g
Specific Gravity 2.01
Solubility Information Solubility in water: immiscible
Formula Weight 345.92
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Perfluorobutyl iodide
Show More Show Less
compliance-icons
Product Identifier
  • Perfluorobutyl iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.