missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(methyldecahydronaphthalene), tech. 85%, mixture of isomers, Thermo Scientific Chemicals

Product Code. 11317625
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317625 100 g 100g
11307625 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11317625 Supplier Thermo Scientific Alfa Aesar Supplier No. L05862.22
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51294-16-7
Molecular Formula C11F20
Molecular Weight (g/mol) 512.09
MDL Number MFCD00014326
InChI Key LWRNQOBXRHWPGE-UHFFFAOYNA-N
Synonym perfluoromethyldecalin, perfluoro methyldecalin, flutec pp9, perfluoro 1-methyldecalin, perfluoro-1-methyldecalin, pp 9, perfluoro methyldecalin , tech., naphthalene, 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8a-heptadecafluorodecahydro-8-trifluoromethyl, heptadecafluorodecahydro-1-trifluoromethyl naphthalene, 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8a-heptadecafluoro-8-trifluoromethyl naphthalene
PubChem CID 39977
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8a-heptadecafluoro-8-(trifluoromethyl)naphthalene
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C12F

Specifications

Melting Point -70°C
Density 1.972
Boiling Point ∼160°C
Refractive Index 1.317
Quantity 100 g
Formula Weight 512.09
Percent Purity 85%
Grade Technical
Chemical Name or Material Perfluoro(methyldecahydronaphthalene), ≈1:1 mixture of 1- and 2-isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.