missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(methylcyclohexane), TRC

Product Code. 30419830
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419830 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419830 Supplier Toronto Research Chemicals Supplier No. TRCP999225250MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluoro(methylcyclohexane)
CAS 355-02-2
Molecular Formula C7F14
Quantity 250 mg
Synonym Cyclohexane','1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)- (ACI), 1,1,2,2,3,3,4,4,5,5,6-Undecafluoro-6-(trifluoromethyl)cyclohexane (ACI), Cyclohexane','undecafluoro(trifluoromethyl)- (6CI','8CI','9CI), Flutec PC 2, Flutec PP 2, Khladon 350, NSC 4779, Perfluoro(methylcyclohexane), PP 2, Undecafluoro(trifluoromethyl)cyclohexane
InChI Formula InChI=1S/C7F14/c8-1(7(19,20)21)2(9,10)4(13,14)6(17,18)5(15,16)3(1,11)12
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)cyclohexane
Molecular Weight (g/mol) 350.05
Formula Weight 349.9776
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.