missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(methylcyclohexane), 94%, Thermo Scientific Chemicals

Product Code. 11442347
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11442347

Brand: Thermo Scientific Alfa Aesar A14233.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for fluorous biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for ″fluorous″ biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.

Solubility
Miscible with acetone and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-02-2
Molecular Formula C7F14
Molecular Weight (g/mol) 350.06
MDL Number MFCD00001468
InChI Key QIROQPWSJUXOJC-UHFFFAOYSA-N
Synonym perfluoro methylcyclohexane, perfluoromethylcyclohexane, flutec pp2, undecafluoro trifluoromethyl cyclohexane, tetradecafluoromethylcyclohexane, cyclohexane, undecafluoro trifluoromethyl, unii-vj9772yw63, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl cyclohexane, cyclohexane, 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-trifluoromethyl, perfluoro methylcyclohexane , pmch
PubChem CID 9637
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)cyclohexane
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F

Specifications

Melting Point -70°C
Density 1.8
Boiling Point 75°C to 76°C
Odor Odorless
Refractive Index 1.3 00
Quantity 100 g
Beilstein 1915981
Solubility Information Miscible with acetone and benzene.
Formula Weight 350.06
Percent Purity 94%
Chemical Name or Material Perfluoro(methylcyclohexane)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.