missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals

Catalog Number 10447320
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
311-89-7
C12F27N
670
MFCD00000436
RVZRBWKZFJCCIB-UHFFFAOYSA-N
perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine
9397
CHEBI:38854
1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
311-89-7
C12F27N
670
MFCD00000436
RVZRBWKZFJCCIB-UHFFFAOYSA-N
perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine
9397
CHEBI:38854
1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -52.0°C
Color Colorless
Density 1.8800g/mL
Boiling Point 173.0°C to 181.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.291
Quantity 10 g
Specific Gravity 1.88
Solubility Information Solubility in water: insoluble.
Viscosity 2.3-3.3 mm2/s (25°C)
Formula Weight 670
Physical Form Liquid
Chemical Name or Material Perfluoro-compound FC-43*
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'