missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(allylbenzene), 97%

Product Code. 11476994
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476994 25 g 25g
11466994 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476994 Supplier Thermo Scientific Alfa Aesar Supplier No. B21469.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 67899-41-6 | C9F10 | 298.083 g/mol

Perfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, ammonia.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67899-41-6
Molecular Formula C9F10
Molecular Weight (g/mol) 298.083
MDL Number MFCD00153225
InChI Key WRHBYJDZKRNITP-UHFFFAOYSA-N
Synonym perfluoro allylbenzene, perfluoroallylbenzene, 3-pentafluorophenyl pentafluoro-1-propene, 3-pentafluorophenyl pentafluoroprop-1-ene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-enyl benzene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-en-1-yl benzene, 1,2,3,4,5-pentafluoro-6-pentafluoroprop-2-en-1-yl benzene, benzene,1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoro-2-propen-1-yl, acmc-1b8oa, 1-pentafluoroallyl pentafluorobenzene
PubChem CID 2775989
IUPAC Name 1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-enyl)benzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(C(=C(F)F)F)(F)F

Specifications

Density 1.708
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 298.08
Percent Purity 97%
Chemical Name or Material Perfluoro(allylbenzene)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.