missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(2-n-butyltetrahydrofuran), tech., cont. perfluoro(2-n-propyltetrahydropyran), Thermo Scientific™

Product Code. 11420611
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420611 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11420611 Supplier Thermo Scientific Alfa Aesar Supplier No. B20098.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a reaction medium with substantial capacity to solubilize molecular oxygen as in the high yield epoxidation of alkenes in the presence of trimethylacetaldehyde.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-36-4
Molecular Formula C8F16O
Molecular Weight (g/mol) 416.06
MDL Number MFCD00014513
InChI Key FYJQJMIEZVMYSD-UHFFFAOYNA-N
Synonym perfluoro-2-butyltetrahydrofuran, perfluoro 2-butyltetrahydrofuran, fluorinert fc-75, perfluoro-compound fc-75, perfluoro butyltetrahydrofuran, heptafluorotetrahydro-2-nonafluorobutyl furan, furan, 2,2,3,3,4,4,5-heptafluorotetrahydro-5-nonafluorobutyl, perfluoro-2-n-butyltetrahydrofurane, 2,2,3,3,4,4,5-heptafluoro-5-nonafluorobutyl tetrahydrofuran, 2,2,3,3,4,4,5-heptafluorotetrahydro-5-nonafluorobutyl furan
PubChem CID 78976
ChEBI CHEBI:39023
IUPAC Name 2,2,3,3,4,4,5-heptafluoro-5-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)oxolane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)OC(F)(F)C(F)(F)C1(F)F

Specifications

Density 1.77
Quantity 250 g
Refractive Index <1.3005
Boiling Point 99°C to 107°C
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 416.06
Grade Technical
Assay cont. perfluoro(2-n-propyltetrahydropyran)
Chemical Name or Material Perfluoro(2-n-butyltetrahydrofuran)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.