missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(2-methyl-3-oxahexanoyl) fluoride, 97%

Product Code. 11309195
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11309195

Brand: Thermo Scientific Alfa Aesar L16483.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2062-98-8 | C6F12O2 | 332.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2062-98-8
Molecular Formula C6F12O2
Molecular Weight (g/mol) 332.045
MDL Number MFCD00054657
InChI Key BCLQALQSEBVVAD-UHFFFAOYSA-N
Synonym 2,3,3,3-tetrafluoro-2-perfluoropropoxy propanoyl fluoride, undecafluoro-2-methyl-3-oxahexanoyl fluoride, perfluoro 2-methyl-3-oxahexanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy propanoyl fluoride, perfluoro-2-methyl-3-oxahexanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy, hfpo dimer, acmc-1cljd
PubChem CID 102740
IUPAC Name 2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoyl fluoride
SMILES C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)F

Specifications

Boiling Point 54°C to 56°C
Flash Point None
Refractive Index 1.3
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 332.06
Percent Purity 97%
Chemical Name or Material Perfluoro(2-methyl-3-oxahexanoyl) fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.