missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride, 97%

Product Code. 11379285
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379285 25 g 25g
11369285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379285 Supplier Thermo Scientific Alfa Aesar Supplier No. L16611.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34761-47-2 | C15F30O5 | 830.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34761-47-2
Molecular Formula C15F30O5
Molecular Weight (g/mol) 830.11
MDL Number MFCD00054660
InChI Key QDHLRGOEEBWEIB-UHFFFAOYNA-N
Synonym perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride, 3,6,9,12-tetraoxapentadecanoyl fluoride, 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis trifluoromethyl, 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis trifluoromethyl-3,6,9,12-tetraoxapentadecanoyl fluoride, acmc-1csyq, 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis trifluoromethyl-3,6,9,12-tetraoxapentadecan-1-oyl fluoride, perfluoro 2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propoxy propanoyl fluoride, 2,5,8,11-tetrakis trifluoromethyl-3,6,9,12-tetraoxaheptadecafluoropentadecanoic acid fluoride, 3,6,9,12-tetraoxapentadecanoylfluoride,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis trifluoromethyl
PubChem CID 118188
SMILES FC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Density 1.8
Boiling Point 200°C to 203°C
Flash Point None
Refractive Index 1.3
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
IUPAC Name 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis(trifluoromethyl)-3,6,9,12-tetraoxapentadecanoyl fluoride
Formula Weight 830.12
Percent Purity 97%
Chemical Name or Material Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.