missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-iodohexane, 98+%, stab. with copper

Product Code. 11470017
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470017 10 g 10g
11480017 50 g 50g
11490017 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470017 Supplier Thermo Scientific Alfa Aesar Supplier No. B20958.09
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 355-43-1 | C6F13I | 445.95 g/mol

It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-43-1
Molecular Formula C6F13I
Molecular Weight (g/mol) 445.95
MDL Number MFCD00001063
InChI Key BULLJMKUVKYZDJ-UHFFFAOYSA-N
Synonym perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo
PubChem CID 67733
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
SMILES C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point -45°C
Density 2.063
Boiling Point 117°C
Refractive Index 1.329
Quantity 10 g
Beilstein 1802118
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 445.95
Percent Purity ≥98%
Chemical Name or Material Perfluoro-1-iodohexane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.