missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-iodohexane, 97%

Product Code. 15492132 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492132 25 g 25g
15482132 5 g 5g
15402142 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492132 Supplier Thermo Scientific Acros Supplier No. 269940250
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 355-43-1 | C6F13I | 445.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-43-1
Molecular Formula C6F13I
Molecular Weight (g/mol) 445.95
InChI Key BULLJMKUVKYZDJ-UHFFFAOYSA-N
Synonym perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo
PubChem CID 67733
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
SMILES C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point -45.0°C
Density 2.0630g/mL
Boiling Point 116.0°C to 119.0°C
Flash Point >115°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CF3(CF2)5I
Refractive Index 1.3270 to 1.3300
Quantity 25 g
Specific Gravity 2.063
Solubility Information Solubility in water: insoluble.
Formula Weight 445.95
Percent Purity 97%
Chemical Name or Material Perfluoro-1-iodohexane
Show More Show Less
compliance-icons
Product Identifier
  • Perfluoro-1-iodohexane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.