missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(1,3-dimethylcyclohexane), tech. 90%, Thermo Scientific Chemicals

Product Code. 15422297
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-27-3
Molecular Formula C8F16
Molecular Weight (g/mol) 400.062
MDL Number MFCD00001469
InChI Key LOQGSOTUHASIHI-UHFFFAOYSA-N
Synonym perfluoro-1,3-dimethylcyclohexane, perfluoro 1,3-dimethylcyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl cyclohexane, decafluoro-1,3-bis trifluoromethyl cyclohexane, cyclohexane, perfluoro-1,3-dimethyl, cyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl, cyclohexane, decafluoro-1,3-bis trifluoromethyl, hexadecafluoro 1,3-dimethylcyclohexane, flutec pp3, acmc-1cjoy
PubChem CID 78975
IUPAC Name 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis(trifluoromethyl)cyclohexane
SMILES C1(C(C(C(C(C1(F)F)(F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F

Specifications

Melting Point -55°C
Density 1.828 g/mL
Boiling Point 101°C to 102°C
Odor Odorless
Quantity 50 g
Beilstein 2064147
Formula Weight 400.06
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Perfluoro (1,3-dimethylcyclohexane)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'