Learn More
Uvex™ Perfexxion Helmet
The Uvex perfexxion safety helmet can be used as an industrial helmet or as a mountaineering or cycling helmet.
POA
Specifications
Certifications/Compliance | EN 1078, EN 397, EN 12492 |
---|---|
Includes | Chin Strap |
Description
- Multi-standard helmet – DIN EN 12492 mountaineering helmet standard, DIN EN 1078 cycling helmet standard and DIN EN 397 industrial safety helmet standard (in part, except 4.4 internal vertical clearance, 4.9 max. ventilation opening cross-section and 5.1.4 chin strap fastening)
- Meets DIN EN 397 optional requirements for increased lateral rigidity LD (lateral deformation) and low temperatures (-30°C)
- Closeable air vents at the front and rear
- Suspension harness with wheel ratchet system for variable width adjustment and height adjustment
- Pre-fitted forked chin strap for EN 12492/EN 1078
Specifications
EN 1078, EN 397, EN 12492 | |
Chin Strap |