Learn More
Honeywell™ PERFECT POLY™ PU First White Gloves
Product Code. 12605333
Change view
Click to view available options
Size:
7
8
9
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12605333 7 Each
12615333 8 Each
12625333 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12605333 Supplier Honeywell™ Supplier No. 213225507
Only null left
Add to basket
Add to basket
Honeywell™ PERFECT POLY™ PU First White Gloves designed for fine handling or for wearing under another glove.
  • 100% Nylon knitted glove
  • White PU coating on the palm and fingers
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and PU coating optimise the glove's breathability
  • Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
  • The PU coating ensures a good grip
  • Non pilling, the polyamide is suitable for precision work
  • The gloves are certified as silicone-free to ensure their use in painting
  • White color enhances the glove's hygiene and cleanliness
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 Basic standard, EN 388-2016 Mechanical risks
Construction Seamless Knit
Description 13 gauge
Coating Material Polyamide, Polyurethane
Color White
Size 7
Length (Metric) 24 cm