Learn More
Honeywell™ PERFECT POLY™ Black Gloves
Click to view available options
Size:
6
7
8
9
Unit Size:
1 pair
Description
- Black polyamide knitted glove
- Gray polyurethane coating on palm and fingertips
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back and PU coating optimise the glove's breathability
- Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
- The PU coating ensures a good grip
- Non pilling, the polyamide is suitable for precision work
- The gloves are certified as silicone-free to ensure their use in painting
- Its dark color makes it more soilproof, thus ensuring a longer life
Specifications
Specifications
| Certifications/Compliance | EN 420-03 Basic standards, EN 388-03 Mechanical hazards |
| Description | 13 gauge |
| Color | Black, Gray |
| Material | Polyamide, Polyurethane |
| Size | 9 |
| Length (Metric) | 25 cm |