missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Perfect Cutting™ Grey Gloves

Product Code. 12675313
Click to view available options
Size:
7
8
9
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12675313

Brand: Honeywell Safety Products™ 213224507

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Handle sharp parts in a dry or slightly greasy environment. Honeywell Safety Products™ Perfect Cutting™ Grey Gloves are knitted for a snug, conforming fit and good dexterity.

  • For handling sharp parts in a dry or slightly greasy environment.
  • Knitted for snug conformity to the hand and good dexterity.
  • Dyneema™ Lycra™ construction for comfort.
  • Elastic composite fiber wrist
  • Ventilated back for breathability.
  • Polyurethane coating on the palm for abrasion resistance.
  • Grey coating allows for work in a slightly dirty environment
  • Do not expose this glove to heat.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CATEGORY II Intermediate risks, EN 420-2003 General requirements, EN 388-2003 Mechanical hazards, EPI category CE 2, Quality assurance ISO 9001/2000, EC certificate number 0075/797/162/01/06/0278, Attestation number 0075/797/162/01/06/0278
Classification Dexterity: 5
Coated Area Palm, Fingertips
Cuff Material Elastic composite fiber
Description Gauge 13
Coating Material Polyurethane
Color Gray
Cut Resistance Rating 3
Material Polyamide, Polyurethane, Lycra, Dyneema™
Product Type Gloves
Size 7
For Use With (Application) Building, Construction, Catering, Energy, Electricity, Glass, Logistics, Metallurgy, Pulp and Paper, Telecom Transportation, Public Service, Automobile, Aviation and Maintenance industries
Puncture Rating 2
Quantity 10Pair/PK
Tear Rating 4
Length (Metric) 23.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.