missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ PerCP-Cy5.5 Beads (CE/IVD)
Description
- Designed for use with FACSComp™ or AutoCOMP™ software and the FACS™ family of flow cytometers (FACSCalibur™, FACSort™, FACScan™, and FACStrak™).
- the beads are used to adjust instrument settings, set fluorescence compensation, and check instrument sensitivity.
- Daily use is recommended for monitoring instrument performance over time. CaliBRITE beads are for in vitro diagnostic use.
Specifications
Specifications
| Concentration | 1.5 x 107 to 2.5 x 107 |
| Content And Storage | Store CaliBRITE beads at 2°C to 8°C and protected from direct light. Do not use after the expiration date shown on the label. Store Bead Dilution Buffer at 2°C to 8°C. |
| For Use With (Application) | Flow Cytometry, Fluorescence Compensation |
| For Use With (Equipment) | Flow cytometer |
| Quantity | 25 Tests |
| Stability | After dilution, CaliBRITE bead suspensions prepared in Bead Dilution Buffer are stable for 8 hours at 2°C to 8°C or for 1 hour at 20°C to 25°C. After dilution, CaliBRITE bead suspensions prepared in FACSFlow sheath fluid are stable for 8 hours at 2°C to 8°C or, if PerCP beads are included, for 1 hour at 2°C to 8°C. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?