missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, Environmental Grade Plus, 65-71%, Thermo Scientific™

Product Code. 15411507
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411507 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15411507 Supplier Thermo Scientific Alfa Aesar Supplier No. 038750.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
CAS 7601-90-3
Melting Point -12°C
Boiling Point 203°C
Density 1.67 g/mL
Odor Odorless
Flash Point 113°C (235°F)
Quantity 500 mL
Molecular Formula ClHO4
Physical Form Liquid, Double Sub-Boiling Quartz Distillation
MDL Number MFCD00011325
UN Number UN1873
Merck Index 14,7153
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES O[Cl](=O)(=O)=O
IUPAC Name perchloric acid
Molecular Weight (g/mol) 100.45
PubChem CID 24247
ChEBI CHEBI:29221
Formula Weight 100.46
Percent Purity 65 to 71%
Grade Environmental Plus
Chemical Name or Material Perchloric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Perchloric acid, Environmental Grade Plus, 65-71%
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P283-Wear fire/flame resistant/retardant clothing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.