missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, Environmental Grade, 67-71%, Thermo Scientific™

Product Code. p-7050590
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 15491497

Brand: Thermo Scientific Alfa Aesar 038749.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Color Colorless
Melting Point -12°C
Boiling Point 203°C
Density 1.67 g/mL
Odor Odorless
Flash Point 113°C (235°F)
Quantity 500 mL
Physical Form Liquid, Single Sub-Boiling Quartz Distillation
UN Number UN1873
Merck Index 14,7153
Formula Weight 100.46
Percent Purity 65 to 71%
Grade Environmental
Chemical Name or Material Perchloric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Perchloric acid, 67-71%
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P283-Wear fire/flame resistant/retardant clothing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.