missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
6 x 500 mL
Unit Size:
1 L
100mL
2.5 L
500mL
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Concentration | 70% |
| Density | 1.664g/mL at 25°C |
| Residue after Ignition | ≤0.003% |
| Vapor Pressure | 6.8mmHg (25°C) |
| Flash Point | 113°C (235.4°F) |
| Quantity | 500 mL |
| Linear Formula | HClO4 |
| UN Number | 1873 |
| Formula Weight | 100.46g/mol |
| Grade | ACS Reagent |
| Show More |
Safety and Handling
Product Identifier
- Perchloric acid
- Danger
- Oxidising liquid Category 1
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- H271-May cause fire or explosion; strong oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : perchloric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction