missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, ≥69% (T), Honeywell™ Fluka™

Product Code. 15677050 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15677050 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15677050 Supplier Honeywell-Fluka Supplier No. 772281L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

pH <1
Color Colorless
CAS 7601-90-3
Melting Point -18°C
Boiling Point 203°C
Density 1.66g/cu.cm at 20°C
Odor Odorless
Packaging Glass bottle
Flash Point 113°C (235.4°F)
Quantity 1 L
Molecular Formula ClHO4
Linear Formula HClO4
Physical Form Liquid
MDL Number MFCD00011325
UN Number UN1873
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
Solubility Information Completely miscible in water
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES O[Cl](=O)(=O)=O
IUPAC Name perchloric acid
Molecular Weight (g/mol) 100.45
PubChem CID 24247
ChEBI CHEBI:29221
Formula Weight 100.46g/mol
Percent Purity ≥69% (T)
Chemical Name or Material Perchloric acid, Puriss. p.a., ACS Reagent, 70% (Hg ≤0.0000005%), packed in coated, shock- and leak-protected glass bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.