missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid 60% d=1.53, Certified AR for Analysis, Fisher Chemical™

Product Code. 15908916 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
500 mL
Packaging:
Amber Glass Bottle
Plastic Coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15908916 1 L Amber Glass Bottle 1 L
10636652 500 mL Plastic Coated Glass Bottle 500mL
10646652 2.5 L Plastic Coated Glass Bottle 2.5 L
12606757 25 L Amber Glass Bottle 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15908916 Supplier Fisher Chemical Supplier No. P/1240/PB15
  • ONLINE EXCLUSIVE PRICE
    £369.00
    £260.00 / 1 L
    Save £109.00
    29% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH 0.1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Packaging Amber Glass Bottle
Vapor Pressure 6.8mmHg 25
Quantity 1 L
Viscosity 3.5 mPaS at 20°C
Physical Form Liquid
Formula Weight 100.46g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.