missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 60%, ACS Reagent, Solstice

Product Code. 15643270
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15643270 100 mL Glass Bottle 100mL
15673270 500 mL Glass Bottle 500mL
15653270 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15643270 Supplier Honeywell-Fluka Supplier No. 311413100ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. Afficher la politique du retour.

ACS Reagent, 60%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Spécification

Melting Point -18°C
Boiling Point 203°C
Density 1.535g/mL at 25°C
Packaging Glass Bottle
Residue after Ignition ≤0.003%
Vapor Pressure 7mmHg (25°C)
Flash Point 113°C (235.4°F)
Quantity 100 mL
Linear Formula HClO4
UN Number UN1873
Formula Weight 100.46g/mol
Percent Purity 60%
Grade ACS Reagent
Chemical Name or Material Perchloric Acid
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.