Learn More
Perchloric acid, 0.1N in Acetic Acid Standardized Solution
Description
Perchloric acid, 0.1N in acetic acid standardized solution is used in volumetric analysis. It is useful for its super acid properties and it is also used as an etchant for certain metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Appearance | Colorless liquid |
| Color | Colorless |
| Odor | Pungent |
| Vapor Pressure | 16 hPa (17mm Hg) at 20°C |
| Quantity | 4 L |
| Physical Form | Liquid |
| Solubility Information | Miscible with water. |
| Formula Weight | 100.46 |
| Chemical Name or Material | Perchloric Acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.