missing translation for 'onlineSavingsMsg'
Learn More

Peramivir trihydrate, MedChemExpress

Product Code. 16459787
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16459787

Brand: Medchem Express HY17015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Peramivir trihydrate (RWJ-270201 trihydrate;BCX-1812 trihydrate) is a highly potent, selective and orally active influenza virus neuraminidase (NA) inhibitor, with IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes.

Peramivir trihydrate (RWJ-270201 trihydrate

Specifications

Chemical Name or Material Peramivir trihydrate
Physical Form Powder
CAS 1041434-82-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H34N4O7
For Use With (Application) COVID-19-anti-virus
Synonym RWJ 270201 trihydrate BCX 1812 trihydrate
Solubility Information H2O : 5 mg/mL (13.07 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMILES O[C@H]1[C@]([C@H](C(CC)CC)NC(C)=O)([H])[C@H](NC(N)=N)C[C@@H]1C(O)=O.[H]O.[H]O.[H]O
Molecular Weight (g/mol) 382.45
Formula Weight 382.45
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.