missing translation for 'onlineSavingsMsg'
Learn More

Peracetic Acid Solution, 0%, Honeywell™ Fluka™

Product Code. 15658350 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15658350 5 mL 5mL
15662820 100 mL 100mL
15672820 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15658350 Supplier Honeywell Research Chemicals Supplier No. 2693365ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 79-21-0
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
MDL Number MFCD00002128
InChI Key KFSLWBXXFJQRDL-UHFFFAOYSA-N
Synonym peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1
PubChem CID 6585
IUPAC Name ethaneperoxoic acid
SMILES CC(=O)OO

Specifications

Density 1.13g/mL at 25°C
Linear Formula CH3CO3H
Quantity 5 mL
Beilstein 1098464
Formula Weight 76.05g/mol
Vapor Pressure 20mmHg (25°C)
Percent Purity 0%
Chemical Name or Material Peracetic Acid Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.