missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PER3 Recombinant Rabbit Monoclonal Antibody (9I3I6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538036
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, Mouse lung, Mouse kidney, Rat lung Immunogen sequence: GRRGAKDEAL GEESGERWSP EFHLQRKLAD SSHSEQQDRN RVSEELIMVV QEMKKYFPSE RRNKPSTLDA LNYALRCVHS VQANSEFFQI L.
This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. This gene is upregulated by CLOCK/ARNTL heterodimers but then represses this upregulation in a feedback loop using PER/CRY heterodimers to interact with CLOCK/ARNTL. Polymorphisms in this gene have been linked to sleep disorders. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| PER3 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70361, P56645, Q8CJE2 | |
| PER3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-100 of human PER3 (P56645). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9I3I6 | |
| Unconjugated | |
| PER3 | |
| 2810049O06Rik; Cell growth-inhibiting gene 13 protein; circadian clock protein PERIOD 3; FASPS3; GIG13; hPER3; mPER3; Per3; period circadian clock 3; period circadian protein 3; period circadian protein homolog 3; period homolog 3; period3; rper3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18628, 78962, 8863 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction