Learn More
Invitrogen™ PER3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA540922
Product Code. 15544492
  • £441.00 / 100µL
Description
Peptide sequence: DATLFCEPWT LNMQPAPLTS EEFKHVGLTA AVLSAHTQKE EQNYVDKFRE Sequence homology: Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 86%; Rat: 93%; Sheep: 86%; Zebrafish: 79%.
This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. This gene is upregulated by CLOCK/ARNTL heterodimers but then represses this upregulation in a feedback loop using PER/CRY heterodimers to interact with CLOCK/ARNTL. Polymorphisms in this gene have been linked to sleep disorders. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PER3
Polyclonal
Unconjugated
PER3
2810049O06Rik; Cell growth-inhibiting gene 13 protein; circadian clock protein PERIOD 3; FASPS3; GIG13; hPER3; mPER3; Per3; period circadian clock 3; period circadian protein 3; period circadian protein homolog 3; period homolog 3; period3; rper3
Rabbit
Affinity Chromatography
RUO
8863
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P56645
PER3
Synthetic peptide directed towards the C-terminal of human PER3 (aa 647-696).