missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PER3 Antibody (SR1135), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | PER3 |
|---|---|
| Clone | SR1135 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613225
|
Novus Biologicals
NBP3-22285-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694265
|
Novus Biologicals
NBP3-22285-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PER3 Monoclonal antibody specifically detects PER3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PER3 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cell growth-inhibiting gene 13 protein, Circadian clock protein PERIOD 3, GIG13, growth-inhibiting protein 13, hPER3, period (Drosophila) homolog 3, period 3, period circadian protein 3, period circadian protein homolog 3, period homolog 3 (Drosophila) | |
| A synthesized peptide derived from human PER3 (Uniprot #: P56645) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1135 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Endocrinology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 8863 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title