Learn More
PER2 (phospho S662) Rabbit anti-Human, Mouse, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PER2.
Brand: Abnova PAB29636.100uL
Description
This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. [provided by RefSeq]
Specifications
| PER2 (phospho S662) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PER2. | |
| In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PER2 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S662 of human PER2. | |
| 100 μL | |
| Primary | |
| PER2 (phospho S662) polyclonal antibody detects endogenous levels of human PER2 only when phosphorylated at serine 662. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| FASPS/KIAA0347 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8864 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.