missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PER1 Recombinant Rabbit Monoclonal Antibody (2Y1M7)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558132

 View more versions of this product

Product Code. 30284562

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MSGPLEGADG GGDPRPGESF CPGGVPSPGP PQHRPCPGPS LADDTDANSN GSSGNESNGH ESRGASQRSS HSSSSGNGKD SALLETTESS KSTNSQSPSP PSSSIAYSLL SASSEQDNPS TSGCSSEQSA RARTQKELMT ALRELKLRLP PERRGKGRSG TLATLQYALA CVKQVQANQE YYQQWSLEEG EPCSMDMSTY.

Circadian rhythmicity is a basic property of phylogenetically diverse organisms which range from animals and plants, to fungi. Regulation of endogenous biological clocks is regulated at the genetic level by a protein-mediated, autoregulatory feed-back loop. In mammals, several genes that encode members of the basic helix-loop helix (bHLH) PAS (PER-ARNT-SIM) transcription factor family have been shown to play a significant role in regulating circadian oscillations. Transactivation of CLOCK-induced genes is mediated via an E box enhancer (CACGTG) found upstream of target genes. CLOCK-ARNT3 heterodimers bind to E box regulatory elements and stimulate gene transcription. CLOCK has been shown to transactivate the mammalian homolog of Drosophila per. PER, in concert with the product of the mammalian timeless gene (TIM), negatively regulates its own transcription by blocking the activity of the CLOCK-BMAL1 transactivation complex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PER1
Recombinant Monoclonal
1.28 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
O15534, O35973
PER1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human PER1 (NP_002607.2).
100 μL
Primary
Human, Mouse
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
2Y1M7
Unconjugated
PER1
circadian clock protein PERIOD 1; Circadian pacemaker protein Rigui; hPER; hPER1; KIAA0482; mPer1; m-rigui; PER; PER1; per1 {ECO:0000312; period 1; period circadian clock 1; period circadian protein homolog 1; period homolog 1; Period, drosophila, homolog of; RGD:727863}; RIGUI; rPER1
Rabbit
Protein A
RUO
18626, 5187
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.