missing translation for 'onlineSavingsMsg'
Learn More

Peptidylglycine alpha-Amidating Monooxygenase/PAM Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18382254 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382254 100 μL 100µL
18302354 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18382254 Supplier Novus Biologicals Supplier No. NBP303010100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptidylglycine alpha-Amidating Monooxygenase/PAM Polyclonal specifically detects Peptidylglycine alpha-Amidating Monooxygenase/PAM in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peptidylglycine alpha-Amidating Monooxygenase/PAM
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias PAL, pancreatic peptidylglycine alpha-amidating monooxygenase, peptidyl alpha-amidating enzyme, peptidyl-alpha-hydroxyglycine alpha-amidating lyase, peptidylamidoglycolate lyase, peptidylglycine 2-hydroxylase, peptidyl-glycine alpha-amidating monooxygenase, peptidylglycine alpha-amidating monooxygenase, peptidylglycine alpha-hydroxylating monooxygenase, PHM
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800 to the C-terminus of human Peptidylglycine alpha-Amidating Monooxygenase/PAM (NP_620176.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5066
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.