missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peptidylglycine alpha-Amidating Monooxygenase/PAM Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Peptidylglycine alpha-Amidating Monooxygenase/PAM Polyclonal specifically detects Peptidylglycine alpha-Amidating Monooxygenase/PAM in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Peptidylglycine alpha-Amidating Monooxygenase/PAM |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | PAL, pancreatic peptidylglycine alpha-amidating monooxygenase, peptidyl alpha-amidating enzyme, peptidyl-alpha-hydroxyglycine alpha-amidating lyase, peptidylamidoglycolate lyase, peptidylglycine 2-hydroxylase, peptidyl-glycine alpha-amidating monooxygenase, peptidylglycine alpha-amidating monooxygenase, peptidylglycine alpha-hydroxylating monooxygenase, PHM |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 800 to the C-terminus of human Peptidylglycine alpha-Amidating Monooxygenase/PAM (NP_620176.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?