missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peptide YY Antibody (4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23584F
This item is not returnable.
View return policy
Description
Peptide YY Monoclonal antibody specifically detects Peptide YY in Human samples. It is validated for Western Blot, ELISA
Specifications
| Peptide YY | |
| Monoclonal | |
| FITC | |
| Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I | |
| Synthetic N-terminal fragment of human PYY(3-36) coupled to carrier. | |
| 0.1 mL | |
| Signal Transduction | |
| 5697 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 4 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction