missing translation for 'onlineSavingsMsg'
Learn More

Peptide YY Antibody (4), DyLight 488, Novus Biologicals™

Product Code. 18430607 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430607 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430607 Supplier Novus Biologicals Supplier No. NBP223584G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide YY Monoclonal antibody specifically detects Peptide YY in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peptide YY
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I
Host Species Mouse
Immunogen Synthetic N-terminal fragment of human PYY(3-36) coupled to carrier.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5697
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.