missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peptidase Inhibitor 16/PI16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£292.00
Specifications
| Antigen | Peptidase Inhibitor 16/PI16 |
|---|---|
| Dilution | Western Blot 0.4 μg/mL, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Peptidase Inhibitor 16/PI16 Polyclonal specifically detects Peptidase Inhibitor 16/PI16 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Peptidase Inhibitor 16/PI16 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CRISP9, CRISP-9, Cysteine-rich secretory protein 9, dJ90K10.5, DKFZp586B1817, MGC45378, microseminoprotein, beta-binding protein, MSMBBP, peptidase inhibitor 16, PI-16, protease inhibitor 16, PSP94-binding protein, PSPBP | |
| PI16 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 μg/mL, Immunohistochemistry | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q6UXB8 | |
| 221476 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CEPIGSPEDAQDLPYLVTEAPSFRATEASDSRKMGTPSSLATGIPAFLVTEVSGSLATKALPAVETQAPTSLATKDPPSMATEAPPCVT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title