missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peptidase Inhibitor 16/PI16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19773
This item is not returnable.
View return policy
Description
Peptidase Inhibitor 16/PI16 Polyclonal specifically detects Peptidase Inhibitor 16/PI16 in Human samples. It is validated for Western Blot.
Specifications
| Peptidase Inhibitor 16/PI16 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| CRISP9, CRISP-9, Cysteine-rich secretory protein 9, dJ90K10.5, DKFZp586B1817, MGC45378, microseminoprotein, beta-binding protein, MSMBBP, peptidase inhibitor 16, PI-16, protease inhibitor 16, PSP94-binding protein, PSPBP | |
| Rabbit | |
| 49 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (81%), Rat (83%), Bovine (87%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.86mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q6UXB8 | |
| PI16 | |
| Recombinant protein encompassing a sequence within the center region of human PI16. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 221476 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction