missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPT1/SLC15A1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93362-0.1ml
This item is not returnable.
View return policy
Description
PEPT1/SLC15A1 Polyclonal antibody specifically detects PEPT1/SLC15A1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| PEPT1/SLC15A1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Caco-2 oligopeptide transporter, HPECT1macrophage oligopeptide transporter PEPT1, HPEPT1, Intestinal H(+)/peptide cotransporter, Oligopeptide transporter, small intestine isoform, PEPT1intestinal H+/peptide cotransporter, Peptide transporter 1, solute carrier family 15 (oligopeptide transporter), member 1, solute carrier family 15 member 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of human SLC15A1 (NP_005064.1). VTGLEFSYSQAPSNMKSVLQAGWLLTVAVGNIIVLIVAGAGQFSKQWAEYILFAALLLVVCVIFAIMARFYTYINPAEIEAQFDEDEKKNRLEKSNPYFMS | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6564 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction