missing translation for 'onlineSavingsMsg'
Learn More

PEPT1/SLC15A1 Antibody (1F8), Novus Biologicals™

Product Code. 18394439 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394439 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394439 Supplier Novus Biologicals Supplier No. H00006564M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PEPT1/SLC15A1 Monoclonal antibody specifically detects PEPT1/SLC15A1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEPT1/SLC15A1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005064
Gene Alias Caco-2 oligopeptide transporter, HPECT1macrophage oligopeptide transporter PEPT1, HPEPT1, Intestinal H(+)/peptide cotransporter, Oligopeptide transporter, small intestine isoform, PEPT1intestinal H+/peptide cotransporter, Peptide transporter 1, solute carrier family 15 (oligopeptide transporter), member 1, solute carrier family 15 member 1
Host Species Mouse
Immunogen SLC15A1 (NP_005064, 473 a.a. ~ 572 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VKDGLNQKPEKGENGIRFVNTFNELITITMSGKVYANISSYNASTYQFFPSGIKGFTISSTEIPPQCQPNFNTFYLEFGSAYTYIVQRKNDSCPEVKVFE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6564
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.